Merck
  • 4(R/S)-Amino/guanidino-substituted proline peptides: design, synthesis and DNA transfection properties.
4(R/S)-Amino/guanidino-substituted proline peptides: design, synthesis and DNA transfection properties.
Chimia (2013-02-12)
M Umashankara, Manaswini Nanda, Mahesh Sonar, Krishna N Ganesh
ABSTRACT
Collagen is a major structural protein found in the connective tissues of higher organisms and mammals and its biomechanical properties are related to the high thermal stability of its triple helical structure. The primary structure of collagen is composed of the repeating tripeptide motif of Pro-Hyp-Gly, where Hyp is 4 R -hydroxy proline. Cationic collagen mimetics consisting of [Pro(X)-Pro(Y)-Gly](6) where Pro(X) and Pro(Y) are 4(R/S)-amino/guanidine proline have been synthesized and shown to form triplexes more stable than the unmodified collagen peptide [Pro-Hyp-Gly](6). The origin of hyperstability is due to conformational pre-organization of proline pucker arising from the electronegativity of the cationic group. These cationic collagen peptides are shown to be effective cell penetrating and plasmid DNA transfecting agents. The results have potential for design of new collagen mimetics for biomaterial applications and efficient cell penetrating agents for drug delivery applications.
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
USP
L-Proline, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
SAFC
L-Proline
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Guanidine sulfate salt, 99%
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Proline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)