Merck
  • Estimation of the enamel and dentin mineral content from the refractive index.
Estimation of the enamel and dentin mineral content from the refractive index.
Caries research (2012-10-12)
I Hariri, A Sadr, S Nakashima, Y Shimada, J Tagami, Y Sumi
ABSTRACT
Recent advances in the field of optics have enabled accurate and localized measurement of optical properties of biological substrates. This work aimed to elucidate the relationship between the local refractive index (n) and mineral content (MC) of enamel and dentin. De- and remineralized lesions in bovine enamel and dentin blocks were sectioned into 300- to 400-µm-thick slices, and placed on a metal plate to capture images of sound, de- and remineralized regions transversely by optical coherence tomography. Mean n at each depth level of the lesion (20- or 40-µm steps for enamel or dentin) was measured by the optical path length-matching method and used to plot n through lesion depth. The specimens were further polished and processed for transverse microradiography for analysis of MC. The n and MC ranged from 1.52 to 1.63 and 50 to 87 (vol.%) in enamel, and from 1.43 to 1.57 and 11 to 48 (vol.%) in dentin, respectively. Strong, positive linear correlations were found between n and MC (Pearson's r = 0.95 and 0.91 for de- and remineralized enamel, and r = 0.94 and 0.91 for dentin, respectively, p < 0.001). Experimental data were validated with a theoretical calculation of n from MC. De- and remineralization of enamel and dentin resulted in measurable changes of n, and, in turn, MC changes of the tissue could be estimated with good accuracy from this long-known optical property by the new analytical approach. Compositional changes of enamel crystallites after remineralization affect n.
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide 0.1 M solution
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium Phosphate Monobasic solution, reagent grade, 1.0 M in solution
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%