Merck
  • Comparison of arylboron-based nucleophiles in Ni-catalyzed Suzuki-Miyaura cross-coupling with aryl mesylates and sulfamates.
Comparison of arylboron-based nucleophiles in Ni-catalyzed Suzuki-Miyaura cross-coupling with aryl mesylates and sulfamates.
The Journal of organic chemistry (2012-06-21)
Na Zhang, David J Hoffman, Nicholas Gutsche, Jayesh Gupta, Virgil Percec
ABSTRACT
The efficiency of arylboron-based nucleophiles, boronic acid, potassium trifluoroborate, neopentylglycolboronate, and pinacol boronate in nickel-catalyzed Suzuki-Miyaura cross-coupling reactions with the two C-O electrophiles, mesylates, and sulfamates was compared. Arylboronic acid is the most reactive and most atom-economic of the four boron species studied. Arylpotassium trifluoroborate cross-couples efficiently only in the presence of water. In the absence of water, aryl neopentylglycolboronate is more efficient, less expensive, and more atom-economic than aryl pinacolboronate.
Product Description
Supelco
Sulfamic acid, analytical standard (for acidimetry), ACS reagent
Sigma-Aldrich
Sulfamic acid, ACS reagent, 99.3%
Sigma-Aldrich
Sulfamic acid, reagent grade, 98%
Sigma-Aldrich
Sulfamic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfamic acid, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfamate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium sulfamate, BioXtra, ≥98.0%
Sigma-Aldrich
Sulfamic acid, ≥99.5% (alkalimetric)