Skip to Content
Merck
All Photos(1)

Documents

07401

Sigma-Aldrich

Sulfamic acid

≥99.5% (alkalimetric)

Synonym(s):

Amidosulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2SO3H
CAS Number:
Molecular Weight:
97.09
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

Assay

≥99.5% (alkalimetric)

form

solid

impurities

≤0.001% heavy metals (as Pb)

ign. residue

≤0.02%

mp

215-225 °C (dec.) (lit.)

density

2.151 g/cm3 at 25 °C

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤1000 mg/kg

cation traces

Fe: ≤30 mg/kg

SMILES string

NS(O)(=O)=O

InChI

1S/H3NO3S/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

IIACRCGMVDHOTQ-UHFFFAOYSA-N

Gene Information

human ... CA1(759) , CA2(760)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfamic acid: a novel and efficient catalyst for the synthesis of aryl-14H-dibenzo [a. j] xanthenes under conventional heating and microwave irradiation.
Rajitha B, et al.
Tetrahedron Letters, 46(50), 8691-8693 (2005)
Solvent effects. 2. Medium effect on the structure, energy, charge density, and vibrational frequencies of sulfamic acid.
Wong MW, et al.
Journal of the American Chemical Society, 114(2), 523-529 (1992)
Sulfamic acid: an efficient, cost-effective and recyclable solid acid catalyst for the Friedlander quinoline synthesis.
Yadav JS, et al.
Tetrahedron Letters, 46(42, 7249-7253 (2005)
Iridium-catalyzed synthesis of primary allylic amines from allylic alcohols: sulfamic acid as ammonia equivalent.
Christian Defieber et al.
Angewandte Chemie (International ed. in English), 46(17), 3139-3143 (2007-03-14)
Sulfamic acid as a cost-effective and recyclable catalyst for liquid Beckmann rearrangement, a green process to produce amides from ketoximes without waste.
Wang B, et al.
Tetrahedron Letters, 45(17), 3369-3372 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service