Skip to Content
Merck
All Photos(2)

Key Documents

A2585

Sigma-Aldrich

Ammonium sulfamate

BioXtra, ≥98.0%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$112.00

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$112.00

About This Item

Linear Formula:
NH4SO3NH2
CAS Number:
Molecular Weight:
114.12
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
powder
solubility:
H2O: 1 M, clear, colorless
water: soluble 114.1 g/L at 20 °C

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥98.0%

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.02% Insoluble matter

ign. residue

≤0.1%

pH

4.5-6.0 (25 °C, 114.1 g/L)

mp

131-135 °C (lit.)

solubility

H2O: 1 M, clear, colorless
water: soluble 114.1 g/L at 20 °C

anion traces

chloride (Cl-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

N.NS(O)(=O)=O

InChI

1S/H3NO3S.H3N/c1-5(2,3)4;/h(H3,1,2,3,4);1H3

InChI key

GEHMBYLTCISYNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium sulfamate (AS) is ammonium salt containing sulfur. It acts as a flame retardant for polyamide 6 (PA6).[1] AS can be prepared by neutralizing sulfamic acid with ammonia. AS is a hygroscopic compound that undergoes decomposition in the soil to form ammonium sulfate. It also shows herbicidal properties.[2]

Application

Ammonium sulfamate may be used to remove unreacted nitrite during quantification of ceftazidime (CFZM) in either pure form or in pharmaceutical preparations by spectrophotometric method.[3] It may be used in the preparation of ammonium dinitramide (ADN).[4]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melnikov NN.
Chemistry of Pesticides., 262-262 (2012)
Flammability of polyamide 6 using the sulfamate system and organo-layered silicate.
Lewin M, et al.
Polymers For Advanced Technologies, 18(9), 737-745 (2007)
Synthesis of Ammonium Dinitramide by Nitration of Potassium and Ammonium Sulfamate. The Effect of Sulfamate Conterion on ADN Purity.
Nazeri GH, et al.
Iranian Journal of Chemistry and Chemical Engineering, 27(1), 85-89 (2008)
Qiu Fang et al.
The Plant cell, 32(12), 3921-3938 (2020-10-23)
Aluminum (Al) is a primary constraint for crop production on acid soils, which make up more than 30% of the arable land in the world. Al resistance in Arabidopsis (Arabidopsis thaliana) is achieved by malate secretion mediated by the Al-ACTIVATED
Basavaraj Hiremath et al.
Acta pharmaceutica (Zagreb, Croatia), 58(3), 275-285 (2008-12-24)
Two spectrophotometric methods for the determination of ceftazidime (CFZM) in either pure form or in its pharmaceutical formulations are described. The first method is based on the reaction of 3-methylbenzothiazolin-2-one hydrazone (MBTH) with ceftazidime in the presence of ferric chloride

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service