Merck
  • Effects of different amine fluoride concentrations on enamel remineralization.
Effects of different amine fluoride concentrations on enamel remineralization.
Journal of dentistry (2012-06-09)
E A Naumova, N Niemann, L Aretz, W H Arnold
ABSTRACT
The aim of this study was to investigate the effects of decreasing fluoride concentrations on repeated demineralizing challenges on human enamel. In 24 teeth, 3mm×3mm windows were prepared on the buccal and lingual sides and treated in a cycling demineralization-remineralization model. Remineralization was achieved with 100, 10 and 0.1 ppm fluoride from anime fluoride. Coronal sections were cut through the artificial lesions, and three sections per tooth were investigated using polarized light microscopy and scanning electron microscopy with quantitative element analysis. The morphology of the lesions was studied, and the extensions of the superficial layer and the body of the lesion were measured. Using element analysis, the Ca, P and F content were determined. The body of the lesion appeared remineralized after application of 100 ppm fluoride, while remineralization of the lesion was less successful after application of 10 and 0.1 ppm fluoride. The thickness of the superficial layer increased with decreasing fluoride concentrations, and also the extension of the body of the lesion increased. Ca and P content increased with increasing fluoride concentrations. The effectiveness of fluoride in enamel remineralization increased with increasing fluoride concentration. A consistently higher level of fluoride in saliva should be a goal in caries prevention.
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydroxyethyl-cellulose, viscosity 90-160 cP, 5 % in H2O(25 °C)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Hydroxyethyl-cellulose
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mw ~380,000
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~720,000
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~1,300,000
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~90,000
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%
Pricing and availability is not currently available.