Merck
  • [(Cp-R)M(CO)3] (M=Re or 99mTc) Arylsulfonamide, arylsulfamide, and arylsulfamate conjugates for selective targeting of human carbonic anhydrase IX.
[(Cp-R)M(CO)3] (M=Re or 99mTc) Arylsulfonamide, arylsulfamide, and arylsulfamate conjugates for selective targeting of human carbonic anhydrase IX.
Angewandte Chemie (International ed. in English) (2012-02-22)
Daniel Can, Bernhard Spingler, Paul Schmutz, Filipa Mendes, Paula Raposinho, Célia Fernandes, Fabrizio Carta, Alessio Innocenti, Isabel Santos, Claudiu T Supuran, Roger Alberto
ABSTRACT
Enhanced receptor selectivity: carbonic anhydrase inhibitors are relevant for both cancer diagnosis and therapy. Combining non-radioactive Re compounds with their radioactive (99m)Tc homologs enables the use of identical molecules for therapy and imaging (theragnostic). The syntheses and in vitro evaluation of [(Cp-R)M(CO)(3)] (Cp=cyclopentadienyl, M=Re, (99m)Tc) with R being a highly potent carbonic-anhydrase-targeting vector is reported.
Sigma-Aldrich
Sulfamic acid, 99.999% trace metals basis
Supelco
Sulfamic acid, analytical standard (for acidimetry), ACS reagent
Sigma-Aldrich
Sulfamic acid, ≥99.5% (alkalimetric)
Sigma-Aldrich
Sulfamic acid, ACS reagent, 99.3%
Sigma-Aldrich
Ammonium sulfamate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium sulfamate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sulfamic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfamic acid, reagent grade, 98%