Merck
  • [The effect of berberine administration of evaluation of the functional state of rat liver after ligation of common bile duct].
[The effect of berberine administration of evaluation of the functional state of rat liver after ligation of common bile duct].
Biomeditsinskaia khimiia (2013-05-09)
I V Zverinskiĭ, N G Mel'nichenko, V A Poplavskiĭ, I P Sut'ko, P G Telegin, A G Shliakhtun
ABSTRACT
On the eighth day after ligation of the common bile duct in rats a significant increase in the serum content of total lipids, cholesterol bilirubin and ALT, alkaline phosphatase, and gamma-glutamyltransferase was observed. In the microsomal fraction there was a marked decrease in the content and activity of microsomal monooxygenases. Introperitoneal injection of berberine (10 mg/kg) for 6 days caused a partial normalization of permeability of hepatocytes plasma membranes and activity microsomal flavin-containing monooxygenases. It is suggested that berberine is a substrate and inducer of flavin-containing monooxygenases. Membrane-stabilizing effect of berberine is probably realized at the level of inhibition of prooxidant status of liver cells.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
γ-Glutamyl Transferase, ERM®, certified reference material
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid
Sigma-Aldrich
γ-Glutamyltranspeptidase from equine kidney, Type VI, 5-12 units/mg solid