Skip to Content
Merck
All Photos(1)

Key Documents

A2237

Sigma-Aldrich

Phosphatase, Alkaline shrimp

≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host

Synonym(s):

Phosphatase, Alkaline from shrimp, Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Sign Into View Organizational & Contract Pricing

Select a Size

500 UNITS
$371.00
1000 UNITS
$678.00

$371.00


Estimated to ship onMay 21, 2025



Select a Size

Change View
500 UNITS
$371.00
1000 UNITS
$678.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$371.00


Estimated to ship onMay 21, 2025


recombinant

expressed in proprietary host

Quality Level

form

buffered aqueous glycerol solution

concentration

≥900 DEA units/mL

foreign activity

RNAse, DNAse, none detected

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Shrimp alkaline phosphatase is a heat labile, hydrolase enzyme. It is a high specific activity alkaline phosphatase purified from a recombinant source. There are two preferred sites for enzyme activity: 5′ protruding, recessive and blunt 5′-termini. The enzyme is irreversibly heat inactivated at 65°C for 15 minutes.

Application

Alkaline phosphatase was used to examine its role in the prevention of high-fat-diet-induced metabolic syndrome in mice.[1] It also may be used in pasteurization process for milks used in dairy products.[2]

Biochem/physiol Actions

Alkaline phosphatase is commonly responsible for dephosphorylation of nucleotides, proteins and alkaloids. The enzyme nonspecifically catalyzes the dephosphorylation of 5′-termini DNA and RNA phosphomonoesters reactions but it does not affect diphosphate or triphosphate linkages.

Unit Definition

One unit will hydrolyze 1 μmole of 4-nitrophenyl phosphate per minute at pH 9.8 at 37 °C.

Physical form

solution in 50% glycerol containing 25 mM Tris-HCl, pH 7.5, 1 mM MgCl2

Analysis Note

Protein determined by biuret

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicoletta I Petridou et al.
Cell, 184(7), 1914-1928 (2021-03-18)
Embryo morphogenesis is impacted by dynamic changes in tissue material properties, which have been proposed to occur via processes akin to phase transitions (PTs). Here, we show that rigidity percolation provides a simple and robust theoretical framework to predict material/structural
Naomi Ofer et al.
The Journal of biological chemistry, 288(16), 11287-11293 (2013-03-09)
Protein degradation via prokaryotic ubiquitin-like protein (Pup) tagging is conserved in bacteria belonging to the phyla Actinobacteria and Nitrospira. The physiological role of this novel proteolytic pathway is not yet clear, although in Mycobacterium tuberculosis, the world's most threatening bacterial
Gouri Chaudhuri et al.
Indian journal of biochemistry & biophysics, 50(1), 64-71 (2013-04-27)
The hydrolysis of p-nitrophenyl phosphate (pNPP) by calf intestinal alkaline phosphatase (CIAP) was investigated with respect to kinetic parameters such as V(max), K(m) and K(cat) under varying pH, buffers, substrate concentration, temperature and period of incubation. Highest activity was obtained
George C Ziobro et al.
Journal of food protection, 76(5), 892-898 (2013-05-07)
Over the past 80 years, a variety of methods have been developed to detect underpasteurized or improperly pasteurized milks used in dairy products. Existing methods are hampered by duration of analysis, poor reproducibility, and in some cases the use of
I V Zverinskiĭ et al.
Biomeditsinskaia khimiia, 59(1), 90-96 (2013-05-09)
On the eighth day after ligation of the common bile duct in rats a significant increase in the serum content of total lipids, cholesterol bilirubin and ALT, alkaline phosphatase, and gamma-glutamyltransferase was observed. In the microsomal fraction there was a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service