Skip to Content
Merck
All Photos(1)

Documents

ERMAD452

γ-Glutamyl Transferase

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®
certified by the BSC

manufacturer/tradename

JRC

format

neat

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMAD452

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gamma-glutamyltransferase predicts functional impairment in elderly adults after ischemic stroke.
Daniele D'Ambrosio et al.
Journal of the American Geriatrics Society, 61(6), 1040-1041 (2013-06-19)
BK virus-associated progressive multifocal leukoencephalopathy.
Kathryn L Daveson et al.
The Medical journal of Australia, 198(4), 216-218 (2013-03-05)
Marco H Bello et al.
Journal of microbiology (Seoul, Korea), 51(1), 88-99 (2013-03-05)
Gamma-glutamyltransferase (GGT, EC 2.3.2.2) cleaves the γ-glutamyl linkage in glutathione (GSH). Ascomycetes in either the Saccharomycotina or the Taphrinomycotina have one to three GGTs, whereas members of the Pezizomycotina have two to four GGTs. A Bayesian analysis indicates there are
Hans-Erik Johansson et al.
Journal of obesity, 2013, 567984-567984 (2013-03-20)
Obesity is characterized by liver steatosis, chronic inflammation, and increased liver enzymes, that is, gamma-glutamyltransferase (GGT) and alanine aminotransferase (ALT), markers for nonalcoholic fatty liver disease (NAFLD) and liver fat content. Increased platelet counts (PCs) are associated with inflammatory conditions
Marc D Samsky et al.
Journal of the American College of Cardiology, 61(24), 2397-2405 (2013-04-23)
Heart failure (HF) is a major public health problem leading to frequent hospitalizations, impaired quality of life, and shortened life expectancy. Heart failure leads to a chronic inability to meet metabolic requirements of end organs or skeletal muscle. Current literature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service