Merck
  • CD95/Fas protects triple negative breast cancer from anti-tumor activity of NK cells.
CD95/Fas protects triple negative breast cancer from anti-tumor activity of NK cells.
iScience (2021-11-25)
Abdul S Qadir, Jean Philippe Guégan, Christophe Ginestier, Assia Chaibi, Alban Bessede, Emmanuelle Charafe-Jauffret, Manon Macario, Vincent Lavoué, Thibault de la Motte Rouge, Calvin Law, Jacob Vilker, Hongbin Wang, Emily Stroup, Matthew J Schipma, Bryan Bridgeman, Andrea E Murmann, Zhe Ji, Patrick Legembre, Marcus E Peter
ABSTRACT
The apoptosis inducing receptor CD95/Fas has multiple tumorigenic activities. In different genetically engineered mouse models tumor-expressed CD95 was shown to be critical for cell growth. Using a combination of immune-deficient and immune-competent mouse models, we now establish that loss of CD95 in metastatic triple negative breast cancer (TNBC) cells prevents tumor growth by modulating the immune landscape. CD95-deficient, but not wild-type, tumors barely grow in an immune-competent environment and show an increase in immune infiltrates into the tumor. This growth reduction is caused by infiltrating NK cells and does not involve T cells or macrophages. In contrast, in immune compromised mice CD95 k.o. cells are not growth inhibited, but they fail to form metastases. In summary, we demonstrate that in addition to its tumor and metastasis promoting activities, CD95 expression by tumor cells can exert immune suppressive activities on NK cells, providing a new target for immune therapy.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for FC, sufficient for 50 assays
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
G 418 disulfate salt solution, 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture