Merck
  • Mechanical Tension Promotes Formation of Gastrulation-like Nodes and Patterns Mesoderm Specification in Human Embryonic Stem Cells.
Mechanical Tension Promotes Formation of Gastrulation-like Nodes and Patterns Mesoderm Specification in Human Embryonic Stem Cells.
Developmental cell (2020-11-19)
Jonathon M Muncie, Nadia M E Ayad, Johnathon N Lakins, Xufeng Xue, Jianping Fu, Valerie M Weaver, Jonathon M Muncie, Nadia M E Ayad, Johnathon N Lakins, Xufeng Xue, Jianping Fu, Valerie M Weaver
ABSTRACT
Embryogenesis is directed by morphogens that induce differentiation within a defined tissue geometry. Tissue organization is mediated by cell-cell and cell-extracellular matrix (ECM) adhesions and is modulated by cell tension and tissue-level forces. Whether cell tension regulates development by modifying morphogen signaling is less clear. Human embryonic stem cells (hESCs) exhibit an intrinsic capacity for self-organization, which motivates their use as a tractable model of early human embryogenesis. We engineered patterned substrates that recapitulate the biophysical properties of the early embryo and mediate the self-organization of "gastrulation-like" nodes in cultured hESCs. Tissue geometries that generated local nodes of high cell-adhesion tension directed the spatial patterning of the BMP4-dependent "gastrulation-like" phenotype by enhancing phosphorylation and junctional release of β-catenin to promote Wnt signaling and mesoderm specification. Furthermore, direct force application via mechanical stretching promoted BMP-dependent mesoderm specification, confirming that tissue-level forces can directly regulate cell fate specification in early human development.
Sigma-Aldrich
Bromobenzene, ≥99.5% (GC)
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~800 by LS, average Mn ~600 by GPC
Sigma-Aldrich
Poly(ethyleneimine) solution, average Mw 1800, 50 wt. % in H2O
Sigma-Aldrich
Denhardt′s Solution 50x, for Northern and Southern blotting, solution
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Di(trimethylolpropane) tetraacrylate, average Mw 466
Sigma-Aldrich
PP1, ≥98% (HPLC)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., Molecular Biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%
Roche
Tth DNA Polymerase, pkg of 500 U (2 x 250_U), sufficient for ≤200 reactions