Skip to Content
Merck
All Photos(3)

Key Documents

216224

Sigma-Aldrich

Potassium persulfate

ACS reagent, ≥99.0%

Synonym(s):

KPS, Potassium peroxodisulfate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$105.00
100 G
$153.00
500 G
$267.00

$105.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$105.00
100 G
$153.00
500 G
$267.00

About This Item

Linear Formula:
K2S2O8
CAS Number:
Molecular Weight:
270.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder or crystals

$105.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

ACS reagent

vapor density

9.3 (vs air)

vapor pressure

<0.1 hPa ( 25 °C)

Assay

≥99.0%

form

powder or crystals

reaction suitability

reagent type: catalyst
reaction type: C-H Activation

reagent type: oxidant

impurities

≤0.001% Cl compounds
≤0.005% insolubles

pH

2.5-4.5 (25 °C, 27 g/L)

cation traces

Fe: ≤5 ppm
Mn: ≤2 ppm
heavy metals (as Pb): ≤0.001%

SMILES string

[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O

InChI

1S/2K.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

USHAGKDGDHPEEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium persulfate is a water-soluble inorganic salt that releases oxygen and exhibits a strongly exothermic reaction. It is stable at room temperature but decomposes at high temperatures. Potassium persulfate is commonly used as an oxidant and in the polymerization of numerous monomers, mainly in the field of hydrogel synthesis. [1]

Application


  • Aggregation-induced electrochemiluminescence enhancement of Ag-MOG for amyloid β 42 sensing.: This study demonstrates the use of potassium persulfate in the aggregation-induced electrochemiluminescence enhancement of Ag-MOG for sensitive detection of amyloid β 42, relevant in Alzheimer′s disease research (Zheng et al., 2023).

  • Optimization of preparation conditions and performance of a new degradable soil water retaining agent.: This research investigates the role of potassium persulfate in optimizing the preparation and performance of a novel degradable soil water-retaining agent, highlighting its potential in agricultural applications (Yumang et al., 2024).

  • A superabsorbent and pH-responsive copolymer-hydrogel based on acemannan from Aloe vera (Aloe barbadensis M.): A smart material for drug delivery.: This article explores the synthesis of a pH-responsive hydrogel incorporating potassium persulfate, demonstrating its application in drug delivery systems (Irfan et al., 2024).

  • Silver-Catalyzed Carbofluorination of Olefins and α-Fluoroolefins with Carbamoyl Radicals.: The study describes a novel silver-catalyzed carbofluorination reaction facilitated by potassium persulfate, offering advancements in synthetic organic chemistry (Vincent & Brioche, 2024).

  • Sustained release delivery of favipiravir through statistically optimized, chemically cross-linked, pH-sensitive, swellable hydrogel.: This research utilizes potassium persulfate in the development of a pH-sensitive hydrogel for the controlled release of favipiravir, enhancing therapeutic efficacy (Khan et al., 2024).

Potassium persulfate is used as an oxidizing agent:

  • in the oxidation of 4-Chloro-3-methylphenol in pressurized hot water and supercritical water in a continuous flow system.[2]
  • in determinations of dissolved organic carbon.[2]
Potassium persulfate (KPS) has also been used as a redox initiator :

  • in the graft polymerization of acrylonitrile onto starch in aqueous solution.[3]
  • in the synthesis of sulfate radical anions via polymerization processes.[4]
  • as polymerization and radical initiator in the synthesis of polyacrylamide-Laponite-lead dioxide nanoparticle hydrogel composites by acrylamide as monomers.[5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics and mechanism of graft polymerization of acrylonitrile onto starch initiated with potassium persulfate
zadeh t mt, et al.
Journal of science (2001)
Potassium persulfate as oxidant in pressurized hot water
Juhani K, et al.
Environmental Science & Technology, 33, 2095-2099 (1999)
Oxidation of 4-chloro-3-methylphenol in pressurized hot water/supercritical water with potassium persulfate as oxidant
Juhani K, et al.
Environmental Science & Technology, 35, 3247-3251 (2001)
A systematic study of triangular silver nanoplates: one-pot green synthesis, chemical stability, and sensing application
Yosia Nico W, et al.
Nanoscale, 9, 11705-11712 (2017)
UV-initiated bubble-free frontal polymerization in aqueous conditions
Paul Michael P, et al.
Macromolecules, 48, 8738-8745 (2015)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service