Merck
Structural and functional roles of ether lipids.
Protein & cell (2017-05-20)
John M Dean, Irfan J Lodhi
ABSTRACT
Ether lipids, such as plasmalogens, are peroxisome-derived glycerophospholipids in which the hydrocarbon chain at the sn-1 position of the glycerol backbone is attached by an ether bond, as opposed to an ester bond in the more common diacyl phospholipids. This seemingly simple biochemical change has profound structural and functional implications. Notably, the tendency of ether lipids to form non-lamellar inverted hexagonal structures in model membranes suggests that they have a role in facilitating membrane fusion processes. Ether lipids are also important for the organization and stability of lipid raft microdomains, cholesterol-rich membrane regions involved in cellular signaling. In addition to their structural roles, a subset of ether lipids are thought to function as endogenous antioxidants, and emerging studies suggest that they are involved in cell differentiation and signaling pathways. Here, we review the biology of ether lipids and their potential significance in human disorders, including neurological diseases, cancer, and metabolic disorders.
Product Description
Avanti
18:1 Diether PC, powder
Avanti
4ME 16:0 Diether DG, Avanti Research - A Croda Brand 999986O
Avanti
18:0 Diether PC, 1,2-di-O-octadecyl-sn-glycero-3-phosphocholine, powder
Avanti
16:0-18:1 Diether PE, Avanti Research - A Croda Brand 999974P, powder
Avanti
16:0-18:1 Diether PC, Avanti Research - A Croda Brand 999983C
Avanti
18:1 Diether PC, 1,2-di-O-(9Z-octadecenyl)-sn-glycero-3-phosphocholine, chloroform
Avanti
4ME 16:0 Diether DG, Avanti Research - A Croda Brand 999986C
Avanti
4ME 16:0 Diether PC, Avanti Research - A Croda Brand 999984C
Avanti
13:0 Diether PC, Avanti Research - A Croda Brand 999988P, powder
Avanti
4ME 16:0 Diether PC, Avanti Research - A Croda Brand
Avanti
06:0 Diether PC, Avanti Research - A Croda Brand 999998P, powder
Avanti
4ME 16:0 Diether PE, Avanti Research - A Croda Brand 999985P, powder