Skip to Content
Merck
All Photos(1)

Documents

999989C

Avanti

18:1 Diether PC

1,2-di-O-(9Z-octadecenyl)-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

Dioctadecenyl PC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H88NO6P
CAS Number:
Molecular Weight:
758.15
UNSPSC Code:
51191904
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (999989C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999989C

concentration

10 mg/mL (999989C-10mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCCCCCCC/C=C\CCCCCCCC)COCCCCCCCC/C=C\CCCCCCCC)=O

General description

18:1 Diether PC (phosphocholine) is a class of glycerophospholipids that has two octadecene chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds. It has choline as the alcohol moiety, attached to the phosphate group.

Application

18:1 Diether PC or 1,2-di-O-(9Z-octadecenyl)-sn-glycero-3-phosphocholine might be used:
  • to prepare proteoliposomes to investigate the effects of ceramide and ceramide phosphate/ for the determination of phospholipase A activity of lecithin-cholesterol acyltransferase (LCAT) in the ether matrix
  • to prepare lipid bilayer membrane to study the influence of glutamic acid residues and pH on the properties of transmembrane helices
  • in large unilamellar vesicles (LUVs) to explore the differential interaction of pH (low) insertion peptide (pHLIP) with it by utilizing fluorescence and circular dichroism (CD) spectroscopic approaches

Biochem/physiol Actions

Lipids prepared using 18:1 Diether PC (phosphocholine), are found to be more stable than the natural ester lipids under alkaline conditions.

Packaging

5 mL Clear Glass Sealed Ampule (999989C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Papasani V Subbaiah et al.
Biochemistry, 45(15), 5029-5038 (2006-04-12)
Sphingomyelin (SM), the second most abundant phospholipid in plasma lipoproteins, was previously shown to be a physiological inhibitor of the lecithin-cholesterol acyltransferase (LCAT) reaction. In this study, we investigated the effects of its metabolites, ceramide and ceramide phosphate, on the
Venkatesan Rajagopalan et al.
Biochimica et biophysica acta. Biomembranes, 1859(3), 484-492 (2017-01-11)
Negatively charged side chains are important for the function of particular ion channels and certain other membrane proteins. To investigate the influence of single glutamic acid side chains on helices that span lipid-bilayer membranes, we have employed GWALP23 (acetyl-GGALW5LALALALALALALW19LAGA-amide) as
Bhagyashree D Rao et al.
Chemistry and physics of lipids, 226, 104849-104849 (2019-12-15)
pH (low) insertion peptide (pHLIP) is a polypeptide from the third transmembrane helix of bacteriorhodopsin. The pH-dependent membrane insertion of pHLIP has been conveniently exploited for translocation of cargo molecules and as a novel imaging agent in cancer biology due
John M Dean et al.
Protein & cell, 9(2), 196-206 (2017-05-20)
Ether lipids, such as plasmalogens, are peroxisome-derived glycerophospholipids in which the hydrocarbon chain at the sn-1 position of the glycerol backbone is attached by an ether bond, as opposed to an ester bond in the more common diacyl phospholipids. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service