Skip to Content
Merck
All Photos(1)

Key Documents

999984P

Avanti

4ME 16:0 Diether PC

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-O-phytanyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$355.00

$355.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$355.00

About This Item

Empirical Formula (Hill Notation):
C48H100NO6P
CAS Number:
Molecular Weight:
818.28
UNSPSC Code:
12352211
NACRES:
NA.25

$355.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (999984P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCC(C)CCCC(C)CCCC(C)CCCC(C)C)COCCC(C)CCCC(C)CCCC(C)CCCC(C)C)=O

InChI key

NMRGXROOSPKRTL-SUJDGPGCSA-N

General description

4ME 16:0 Diether PC or 1,2-di-O-phytanyl-sn-glycero-3-phosphocholine is an archaeal phospholipid[1] having two phytane chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds. It has choline as the alcohol moiety, attached to the phosphate group.[2]

Application

4ME 16:0 Diether PC or 1,2-di-O-phytanyl-sn-glycero-3-phosphocholine is suitable for the preparation of multilamellar and giant lipid vesicles.[1] It has also been used as a standard to analyze and compare the archaeal abundances in microbial communities based on the phosphoether lipids (PEL) concentrations.[3]

Biochem/physiol Actions

Ether lipids are components of membrane structure, influencing the fluidity and fusion of biological membranes. They possess antioxidative action and helps reducing oxidative stress. Cell differentiation and cellular signalling are some of the other physiological processes controlled by ether lipids.[2]

Packaging

5 mL Clear Glass Sealed Ampule (999984P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ZhangFei Su et al.
Langmuir : the ACS journal of surfaces and colloids, 34(21), 6249-6260 (2018-05-04)
The insertion and ion-conducting channel properties of alamethicin reconstituted into a 1,2-di- O-phytanyl- sn-glycero-3-phosphocholine bilayer floating on the surface of a gold (111) electrode modified with a 1-thio-β-d-glucose (β-Tg) self-assembled monolayer were investigated using a combination of electrochemical impedance spectroscopy
S A Carr et al.
Geobiology, 11(4), 377-395 (2013-05-21)
Marine sediments of the Ross Sea, Antarctica, harbor microbial communities that play a significant role in the decomposition, mineralization, and recycling of organic carbon (OC). In this study, the cell densities within a 153-cm sediment core from the Ross Sea
Marta Salvador-Castell et al.
Biochimica et biophysica acta. Biomembranes, 1862(2), 183130-183130 (2019-11-18)
It is now well established that cell membranes are much more than a barrier that separate the cytoplasm from the outside world. Regarding membrane's lipids and their self-assembling, the system is highly complex, for example, the cell membrane needs to
Vid Šuštar et al.
PloS one, 7(7), e39401-e39401 (2012-07-14)
As variance from standard phospholipids of eubacteria and eukaryotes, archaebacterial diether phospholipids contain branched alcohol chains (phytanol) linked to glycerol exclusively with ether bonds. Giant vesicles (GVs) constituted of different species of archaebacterial diether phospholipids and glycolipids (archaeosomes) were prepared
John M Dean et al.
Protein & cell, 9(2), 196-206 (2017-05-20)
Ether lipids, such as plasmalogens, are peroxisome-derived glycerophospholipids in which the hydrocarbon chain at the sn-1 position of the glycerol backbone is attached by an ether bond, as opposed to an ester bond in the more common diacyl phospholipids. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service