Merck
  • A widely distributed metalloenzyme class enables gut microbial metabolism of host- and diet-derived catechols.
A widely distributed metalloenzyme class enables gut microbial metabolism of host- and diet-derived catechols.
eLife (2020-02-19)
Vayu Maini Rekdal, Paola Nol Bernadino, Michael U Luescher, Sina Kiamehr, Chip Le, Jordan E Bisanz, Peter J Turnbaugh, Elizabeth N Bess, Emily P Balskus
ABSTRACT
Catechol dehydroxylation is a central chemical transformation in the gut microbial metabolism of plant- and host-derived small molecules. However, the molecular basis for this transformation and its distribution among gut microorganisms are poorly understood. Here, we characterize a molybdenum-dependent enzyme from the human gut bacterium Eggerthella lenta that dehydroxylates catecholamine neurotransmitters. Our findings suggest that this activity enables E. lenta to use dopamine as an electron acceptor. We also identify candidate dehydroxylases that metabolize additional host- and plant-derived catechols. These dehydroxylases belong to a distinct group of largely uncharacterized molybdenum-dependent enzymes that likely mediate primary and secondary metabolism in multiple environments. Finally, we observe catechol dehydroxylation in the gut microbiotas of diverse mammals, confirming the presence of this chemistry in habitats beyond the human gut. These results suggest that the chemical strategies that mediate metabolism and interactions in the human gut are relevant to a broad range of species and habitats.
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
(±)-Epinephrine hydrochloride
Sigma-Aldrich
(+)-Catechin hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
6-Hydroxydopamine hydrochloride, ≥97% (titration), powder
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sigma-Aldrich
DL-3,4-Dihydroxymandelic acid, 95%
Supelco
Diquat dibromide monohydrate, PESTANAL®, analytical standard
Sigma-Aldrich
Benzyl viologen dichloride, 97%
Sigma-Aldrich
Sodium hydrosulfite, technical grade
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins
Sigma-Aldrich
Tyramine hydrochloride, ≥98%
Sigma-Aldrich
3-Hydroxyphenylacetic acid, ≥99%
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Methyl viologen dichloride hydrate, 98%
Sigma-Aldrich
3-Methoxytyramine hydrochloride, ≥95.5%, crystalline
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Enterobactin, from Escherichia coli, ≥98% (HPLC)
Supelco
Carbidopa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium molybdate, ≥98%