Merck
Cellular and Molecular Probing of Intact Human Organs.
Cell (2020-02-16)
Shan Zhao, Mihail Ivilinov Todorov, Ruiyao Cai, Rami Ai -Maskari, Hanno Steinke, Elisabeth Kemter, Hongcheng Mai, Zhouyi Rong, Martin Warmer, Karen Stanic, Oliver Schoppe, Johannes Christian Paetzold, Benno Gesierich, Milagros N Wong, Tobias B Huber, Marco Duering, Oliver Thomas Bruns, Bjoern Menze, Jan Lipfert, Victor G Puelles, Eckhard Wolf, Ingo Bechmann, Ali Ertürk
ABSTRACT
Optical tissue transparency permits scalable cellular and molecular investigation of complex tissues in 3D. Adult human organs are particularly challenging to render transparent because of the accumulation of dense and sturdy molecules in decades-aged tissues. To overcome these challenges, we developed SHANEL, a method based on a new tissue permeabilization approach to clear and label stiff human organs. We used SHANEL to render the intact adult human brain and kidney transparent and perform 3D histology with antibodies and dyes in centimeters-depth. Thereby, we revealed structural details of the intact human eye, human thyroid, human kidney, and transgenic pig pancreas at the cellular resolution. Furthermore, we developed a deep learning pipeline to analyze millions of cells in cleared human brain tissues within hours with standard lab computers. Overall, SHANEL is a robust and unbiased technology to chart the cellular and molecular architecture of large intact mammalian organs.
Sigma-Aldrich
Methyl-β-cyclodextrin, average Mn 1310
Sigma-Aldrich
Methyl salicylate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Eukitt® Quick-hardening mounting medium, for microscopy
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Cresyl Violet acetate, certified by the BSC
Sigma-Aldrich
Dibenzyl ether, purum, ≥98.0% (GC)
Sigma-Aldrich
N-Methyldiethanolamine, ≥99%
Sigma-Aldrich
trans-1-Acetyl-4-hydroxy-L-proline, ≥98%
Sigma-Aldrich
trans-Cinnamaldehyde, 97%
Sigma-Aldrich
Benzyl benzoate, ≥99%, FCC, FG
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile