Skip to Content
Merck
All Photos(1)

Documents

W213802

Sigma-Aldrich

Benzyl benzoate

≥99%, FCC, FG

Synonym(s):

Benzoic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH2C6H5
CAS Number:
Molecular Weight:
212.24
FEMA Number:
2138
Beilstein:
2049280
EC Number:
Council of Europe no.:
262
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.727
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515
FDA 21 CFR 175.105

vapor pressure

1 mmHg ( 125 °C)

Assay

≥99%

form

liquid

autoignition temp.

896 °F

refractive index

n20/D 1.568 (lit.)

bp

323-324 °C (lit.)

mp

17-20 °C (lit.)

density

1.118 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; balsamic; floral; sweet

SMILES string

O=C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

SESFRYSPDFLNCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benzyl benzoate is an aromatic ester that is used as a food flavoring agent. It has been identified as one of the main volatile aroma component of cranberry, mango and Egyptian Jasminum sambac flowers.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yannai S
Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients (2012)
Isolation and characterization of the volatile aroma compounds from the concrete headspace and the absolute of Jasminum sambac (L.) Ait.(Oleaceae) flowers grown in Egypt
Edris AE, et al
European Food Research and Technology, 226(3), 621-621 (2008)
Biosynthesis of benzaldehyde, benzyl alcohol and benzyl benzoate from benzoic acid in cranberry (Vaccinium macrocarpon)
Croteau R
Journal of Food Biochemistry, 1(4), 317-326 (1978)
Thomas S Lisse et al.
NPJ Regenerative medicine, 5, 13-13 (2020-06-23)
Glial-cell-derived neurotrophic factor (GDNF) is a well-studied neuroregenerative factor; however, the degree to which it supports hair formation and skin wound repair is not known. By using a Gfra1 (GDNF family receptor alpha 1) knock-in reporter mouse line, GDNF signaling
Contribution of volatile compounds to mango (Mangifera indica L.) aroma
Pino JA & Mesa J
Flavour and Fragrance Journal, 21(2), 207-213 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service