Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Doped graphene
Linear Formula:
C
x
N
y
P
z
Products
Brand
Sigma-Aldrich (6)
Physical Form
Purity
Chemical Composition
Quality Segment
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
900527
Doped graphene,
nitrogen-doped, avg. no. of layers, 1 ‑ 5
Expand
View Pricing
900526
Doped graphene,
boron-doped, avg. no. of layers, 1 ‑ 5
Expand
View Pricing
900529
Doped graphene,
phosphorus-doped, avg. no. of layers, 1 ‑ 5
Expand
View Pricing
900528
Doped graphene,
sulfur-doped, avg. no. of layers, 1 ‑ 5
Expand
View Pricing
900531
Doped graphene,
nitrogen/phosphorus co-doped, avg. no. of layers, 1 ‑ 5
Expand
View Pricing
900530
Doped graphene,
nitrogen/sulfur co-doped, avg. no. of layers, 1 ‑ 5
Expand
View Pricing
900535
Doped graphene,
boron/nitrogen-doped, avg. no. of layers, 1 ‑ 5
Expand
View Pricing