Skip to Content
Merck
All Photos(1)

Key Documents

900529

Sigma-Aldrich

Doped graphene

phosphorus-doped, avg. no. of layers, 1 ‑ 5

Synonym(s):

P-Doped graphene

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$466.00

$466.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$466.00

About This Item

UNSPSC Code:
12141908
NACRES:
NA.23

$466.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Phosphorus-doped graphene, avg. no. of layers, 1 ‑ 5

Assay

≥95%

Quality Level

form

powder

feature

avg. no. of layers 1 ‑ 5

composition

Carbon, 85-95%
Oxygen, <7.5%
Phosphorus, 1.0-3.0%

surface area

>500 m2/g , BET

General description

Typical thickness: 1-5 layers.
Typical size : 0.5-5 μm.

Application

  • Electrocatalyst.
  • Field-effect transistors.
  • Sensors.
  • Lithium ion batteries.
  • Supercapacitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorus-doped graphene nanosheets as efficient metal-free oxygen reduction electrocatalysts.
Li R, et al.
Royal Society of Chemistry Advances, 3(25), 9978-9984 (2013)
Chenzhen Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 25(35), 4932-4937 (2013-07-19)
We develop a simple and economical thermal annealing method for the synthesis of phosphorus-doped graphene, which exhibits remarkable electrocatalytic activity towards the oxygen reduction reaction and enhances the electrochemical performance as an anode material for lithium ion batteries. The experimental

Articles

Advances in scalable synthesis and processing of two-dimensional materials

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service