Skip to Content
Merck
All Photos(1)

Key Documents

900530

Sigma-Aldrich

Doped graphene

nitrogen/sulfur co-doped, avg. no. of layers, 1 ‑ 5

Synonym(s):

N/S co-doped graphene, NSG, S/N co-doped graphene, SNG

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$451.00

$451.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$451.00

About This Item

UNSPSC Code:
12141908
NACRES:
NA.23

$451.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Nitrogen/Sulfur co-doped graphene, avg. no. of layers, 1 ‑ 5

Assay

≥95%

Quality Level

form

powder

feature

avg. no. of layers 1 ‑ 5

composition

Carbon, 85-95%
Nitrogen, 2.0-4.0%
Oxygen, <7.5%
Sulfur, 2.0-4.0%

surface area

>500 m2/g , BET

General description

Typical thickness: 1-5 layers.
Typical size : 0.5-5 μm.

Application

  • Electrocatalyst.
  • Field-effect transistors.
  • Sensors.
  • Lithium ion batteries.
  • Supercapacitors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfur and nitrogen dual-doped mesoporous graphene electrocatalyst for oxygen reduction with synergistically enhanced performance.
Ji Liang et al.
Angewandte Chemie (International ed. in English), 51(46), 11496-11500 (2012-10-12)
Nitrogen, sulfur-codoped graphene sponge as electroactive carbon interlayer for high-energy and -power lithium-sulfur batteries.
Xing LB, et al.
Journal of Power Sources, 303, 22-28 (2016)
Dan Qu et al.
Nanoscale, 5(24), 12272-12277 (2013-10-24)
A facile hydrothermal synthesis route to N and S, N co-doped graphene quantum dots (GQDs) was developed by using citric acid as the C source and urea or thiourea as N and S sources. Both N and S, N doped

Articles

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Advances in scalable synthesis and processing of two-dimensional materials

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service