All Photos(3)
About This Item
Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
Recommended Products
grade
reagent grade
product line
Vetec™
SMILES string
O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2[C@H](O[C@@H]([C@@H]([C@H]2O)O)O)CO
InChI
1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5-,6+,7-,8-,9-,10-,11+,12-/m1/s1
InChI key
GUBGYTABKSRVRQ-ASMJPISFSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J Jane et al.
Carbohydrate research, 132(1), 105-118 (1984-09-01)
Human-salivary, porcine-pancreatic, and Bacillus subtilis alpha amylases were used to study the structure of amylose-V complexes with butyl alcohol, tert-butyl alcohol, 1,1,2,2-tetrachloroethane, and 1-naphthol, and of retrograded amylose. Alpha amylase hydrolyzes the amorphous, folding areas on the surfaces of the
Amy Hui-Mei Lin et al.
Journal of agricultural and food chemistry, 59(11), 5890-5898 (2011-05-11)
Branch chain-length distribution of amylopectin plays an important role on the characteristics of starch. One of the adapted protocols for determining the chain-length distribution and mass proportion of starch molecules is that starch is debranched with isoamylase and then analyzed
G H Te Wierik et al.
Pharmaceutical research, 11(1), 108-110 (1994-01-01)
Drug release from fatty suppository bases containing a solid dispersion of diazepam with amylodextrin or a complex of prednisolone with amylodextrin was analyzed in a flow-through model. Being present as a suspension in the fatty base, particles of complex or
Yongfeng Ai et al.
Carbohydrate polymers, 92(1), 120-127 (2012-12-12)
This study aimed to understand effects of lipids, including corn oil (CO), soy lecithin (SL), palmitic acid (PA), stearic acid (SA), oleic acid (OA), and linoleic acid (LA), on the enzymatic hydrolysis and physical properties of normal corn (NCS), tapioca
R Steendam et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 14(3), 245-254 (2001-09-29)
Amylodextrin, a starch-based controlled release excipient, spontaneously absorbs moisture during storage. The aim of this study was to investigate plasticisation of amylodextrin by moisture and its effect on compaction and tablet characteristics. The glass transition temperature (T(g)) of amylodextrin powders
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service