Skip to Content
Merck
All Photos(2)

Key Documents

V900421

Sigma-Aldrich

Uridine

Vetec, reagent grade, 99%

Synonym(s):

1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O6
CAS Number:
Molecular Weight:
244.20
Beilstein:
754902
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

mp

163-167 °C (lit.)

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=O)NC2=O

InChI

1S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15H,3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1

InChI key

DRTQHJPVMGBUCF-XVFCMESISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Uridine is a pyrimidine nucleoside composed of uracil and ribose that is readily taken up by the brain.[1] This compound is synthesized de novo in mammals. It is found circulating in the blood and cerebrospinal fluid.[2]

Biochem/physiol Actions

Uridine is an essential compound involved in maintaining cellular function and energy metabolism. It aids in multiple biological processes, including RNA synthesis, the synthesis of biomembranes, and glycosylation.[3] Furthermore, uridine serves as a precursor molecule for UDP-glucose, a vital component in the synthesis and storage of glycogen in the liver.[4] In addition, studies have shown that uridine reduces cytotoxicity and improves neurophysiological functions.[2] It is involved in regulating various normal physiological processes in the body, including the cardio-circulatory, reproductive, peripheral and central nervous, and respiratory systems.[5]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian E Crandall et al.
Journal of medicinal chemistry, 56(6), 2348-2358 (2013-02-16)
Resistance by Plasmodium falciparum to almost all clinically used antimalarial drugs requires the development of new classes of antimalarials. 6-Iodouridine (15), a novel and potent inhibitor of orotidine 5'-monophosphate decarboxylase (ODCase), exhibited efficacy in a mouse model infected by P.
Laure Jobert et al.
Molecular cell, 49(2), 339-345 (2012-12-19)
Single-strand-selective monofunctional uracil-DNA glycosylase 1 (SMUG1) is a base excision repair enzyme that removes uracil and oxidised pyrimidines from DNA. We show that SMUG1 interacts with the pseudouridine synthase Dyskerin (DKC1) and colocalizes with DKC1 in nucleoli and Cajal bodies.
G P Connolly et al.
Trends in pharmacological sciences, 20(5), 218-225 (1999-06-04)
There are many disorders of pyrimidine metabolism and those that involve an alteration in uridine metabolism have neurological and systemic effects, which provide insights into the biological activity of uridine and its analogues. Studies of the metabolism and actions of
Yuan Zhou et al.
Nucleic acids research, 41(13), 6664-6673 (2013-05-10)
Triplex is emerging as an important RNA tertiary structure motif, in which consecutive non-canonical base pairs form between a duplex and a third strand. RNA duplex region is also often functionally important site for protein binding. Thus, triplex-forming oligonucleotides (TFOs)
Che C Colpitts et al.
Journal of virology, 87(7), 3640-3654 (2013-01-04)
Entry of enveloped viruses requires fusion of viral and cellular membranes. Fusion requires the formation of an intermediate stalk structure, in which only the outer leaflets are fused. The stalk structure, in turn, requires the lipid bilayer of the envelope

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service