Skip to Content
Merck
All Photos(1)

Key Documents

V900136

Sigma-Aldrich

Lactic acid solution

Vetec, reagent grade, 85%

Synonym(s):

2-Hydroxypropionic acid, DL-Lactic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CO2H
CAS Number:
Molecular Weight:
90.08
Beilstein:
5238667
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

85%

form

viscous liquid

SMILES string

CC(O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains varying amounts of intermolecular esterification products.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stephen Yiu Chuen Choi et al.
The Journal of pathology, 230(4), 350-355 (2013-06-05)
The common preference of cancers for lactic acid-generating metabolic energy pathways has led to proposals that their reprogrammed metabolism confers growth advantages such as decreased susceptibility to hypoxic stress. Recent observations, however, suggest that it generates a novel way for
Annabelle Olsson et al.
Journal of wildlife diseases, 49(3), 560-567 (2013-06-20)
Using a prospective, randomized study design we demonstrate that midazolam sedation minimizes acidosis compared with physical restraint in captive juvenile estuarine crocodiles during handling or noninvasive procedures at preferred body temperature. A dose of midazolam (5.0 mg/kg) was administered intramuscularly
Paula Gaspar et al.
Biotechnology advances, 31(6), 764-788 (2013-04-10)
The lactic acid bacteria (LAB) are a functionally related group of low-GC Gram-positive bacteria known essentially for their roles in bioprocessing of foods and animal feeds. Due to extensive industrial use and enormous economical value, LAB have been intensively studied
Mohamed Ali Abdel-Rahman et al.
Biotechnology advances, 31(6), 877-902 (2013-04-30)
Fermentative production of optically pure lactic acid has roused interest among researchers in recent years due to its high potential for applications in a wide range of fields. More specifically, the sharp increase in manufacturing of biodegradable polylactic acid (PLA)
Michael Hawkes et al.
The American journal of tropical medicine and hygiene, 90(4), 605-608 (2014-03-05)
Severe malaria is frequently managed without access to laboratory testing. We report on the performance of point-of-care tests used to guide the management of a cohort of 179 children with severe malaria in a resource-limited Ugandan hospital. Correlation coefficients between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service