Skip to Content
Merck
All Photos(1)

Key Documents

V800167

Sigma-Aldrich

D-Mannitol

AR, ≥99%

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

grade

AR

product line

Vetec

Assay

≥99%

mp

167-170 °C (lit.)

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heikki O Koskela et al.
Chest, 124(4), 1341-1349 (2003-10-14)
To compare the sensitivity and validity of mannitol, histamine, and cold air challenges to demonstrate the effect of inhaled corticosteroids (ICS) in asthma. A prospective study. Seventeen patients with recently diagnosed, steroid-naive asthma who fulfilled the diagnostic criteria of Finnish
Maria Badal Tejedor et al.
International journal of pharmaceutics, 486(1-2), 315-323 (2015-04-07)
Tablets are the most convenient form for drug administration. However, despite the ease of manufacturing problems such as powder adhesion occur during the production process. This study presents surface and structural characterization of tablets formulated with commonly used excipients (microcrystalline
Traumatic intracranial hypertension.
Nino Stocchetti et al.
The New England journal of medicine, 370(22), 2121-2130 (2014-05-30)
Cecilio F Caldeira et al.
Nature communications, 5, 5365-5365 (2014-11-06)
Circadian rhythms enable plants to anticipate daily environmental variations, resulting in growth oscillations under continuous light. Because plants daily transpire up to 200% of their water content, their water status oscillates from favourable during the night to unfavourable during the
Diana Bilton et al.
Thorax, 69(12), 1073-1079 (2014-09-24)
Bronchiectasis is characterised by excessive production of mucus and pulmonary exacerbations. Inhaled osmotic agents may enhance mucociliary clearance, but few long-term clinical trials have been conducted. To determine the impact of inhaled mannitol on exacerbation rates in patients with non-cystic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service