![D-Mannitol NIST® SRM® 920](/deepweb/assets/sigmaaldrich/product/structures/357/379/34dd2590-b818-4039-b6dc-b769949ee977/640/34dd2590-b818-4039-b6dc-b769949ee977.png)
D-Mannitol
![D-Mannitol NIST® SRM® 920](/deepweb/assets/sigmaaldrich/product/structures/357/379/34dd2590-b818-4039-b6dc-b769949ee977/640/34dd2590-b818-4039-b6dc-b769949ee977.png)
Synonym(s):
Mannite, D-Mannitol
Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Products
Product Category
Brand
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Special Grade
Showing 1-16 of 16
Compare
Product Number
Description
Pricing