Skip to Content
Merck
All Photos(1)

Key Documents

1720600

USP

Xylitol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Xylite

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$590.00

$590.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$590.00

About This Item

Linear Formula:
HOCH2[CH(OH)]3CH2OH
CAS Number:
Molecular Weight:
152.15
Beilstein:
1720523
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$590.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

xylitol

manufacturer/tradename

USP

mp

94-97 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4+,5+

InChI key

HEBKCHPVOIAQTA-SCDXWVJYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Xylitol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morihiro Fujimura et al.
The Journal of pharmacology and experimental therapeutics, 350(3), 543-551 (2014-06-15)
We characterized TRK-130 (N-[(5R,6R,14S)-17-(cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxymorphinan-6-yl]phthalimide; naltalimide), an opioid ligand, to clarify the therapeutic potential for overactive bladder (OAB). In radioligand-binding assays with cells expressing human µ-opioid receptors (MORs), δ-opioid receptors (DORs), or κ-opioid receptors (KORs), TRK-130 showed high selectivity for MORs
Liang Wei et al.
Bioresource technology, 187, 246-254 (2015-04-12)
In this work, Scheffersomyces stipitis, the yeast with excellent xylose-utilizing ability, was firstly engineered for fumaric acid production from xylose with the heterologous reductive pathway from Rhizopus oryzae FM19, and 1.86g/L fumaric acid was produced by the initial strain PSRPMF
S Lakio et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 90, 16-21 (2014-12-03)
Tablet compression process has been studied over the years from various perspectives. However what exactly happens to material during compression is still unknown. In this study a novel compression die which enables real-time spectroscopic measurements during the compression of material
Leshuai W Zhang et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 78, 105-115 (2015-02-11)
This research aimed to develop in vitro methods to assess hazard of canine food ingredients. Canine hepatocytes were harvested and cell viability of clove-leaf oil (CLO), eugenol (EUG), lemongrass oil (LGO), guanosine monophosphate (GMP), inosine monophosphate (IMP), sorbose, ginger-root extract
Sachin Kumar et al.
Bioprocess and biosystems engineering, 38(1), 39-47 (2014-08-06)
Fermentation of xylose-rich and glucose-rich bagasse hydrolysates, obtained from the two-stage acid hydrolysis was studied using the thermotolerant yeast Kluyveromyces sp. IIPE453. The yeast could grow on xylose-rich hydrolysate at 50 °C with the dry cell weight, cell mass yield and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service