Merck
Spectroscopic insight for tablet compression.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-12-03)
S Lakio, H Ylinärä, O Antikainen, H Räikkönen, J Yliruusi
ABSTRACT
Tablet compression process has been studied over the years from various perspectives. However what exactly happens to material during compression is still unknown. In this study a novel compression die which enables real-time spectroscopic measurements during the compression of material is represented. Both near infrared and Raman spectroscope probes can be attached to the die. In this study the usage of the die is demonstrated by using Raman spectroscopy. Eicosane, d-glucose anhydrate, α-lactose monohydrate and xylitol were used in the study because their compression behavior and bonding properties during compression were assumed to be different. The intensity of the Raman signal changed during compression with all of the materials. However, the intensity changes were different within the materials. The biggest differences were within the xylitol spectra. It was noticed that some peaks disappeared with higher compression pressures indicating that the pressure affected variously on different bonds in xylitol structure. These reversible changes were supposed to relate the changes in conformation and crystal structure. As a conclusion, the die was found to be a significant addition for studying compression process in real-time. It can help to reveal Process induced transformations (PITs) occurring during powder compaction.
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Supelco
Eicosane, analytical standard
Sigma-Aldrich
Eicosane, 99%
Sigma-Aldrich
Xylitol, ≥99% (GC)
Sigma-Aldrich
Xylitol
USP
Xylitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Xylitol, European Pharmacopoeia (EP) Reference Standard