Skip to Content
Merck
All Photos(1)

Key Documents

1612619

USP

N-Acetylneuraminic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-Acetylneuraminic acid dihydrate, o-Sialic acid

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$633.00

$633.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
200 MG
$633.00

About This Item

Empirical Formula (Hill Notation):
C11H19NO9 · 2H2O
CAS Number:
Molecular Weight:
345.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$633.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

n-acetylneuraminic acid

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-65 to -96°C

SMILES string

N([C@H]1[C@@H](O[C@](C[C@@H]1O)(O)C(=O)O)[C@H](O)[C@H](O)CO)C(=O)C

InChI

1S/C11H19NO9/c1-4(14)12-7-5(15)2-11(20,10(18)19)21-9(7)8(17)6(16)3-13/h5-9,13,15-17,20H,2-3H2,1H3,(H,12,14)(H,18,19)/t5-,6+,7+,8+,9+,11-/m0/s1

InChI key

SQVRNKJHWKZAKO-PFQGKNLYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
N-Acetylneuraminic Acid is a commonly determined monosaccharide formed as a degradation product of therapeutic glycoproteins.

Application

N-Acetylneuraminic acid USP reference standard for use in specified quality tests and assays.

Also used to prepare standard stock solution during oligosaccharide and sialic acid analysis according to the given general chapters of United States Pharmacopeia (USP):
  • 〈210〉 Monosaccharide analysis [1]
  • 〈129〉 Analytical procedures for recombinant therapeutic monoclonal antibodies [2]

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical Procedures for Recombinant Therapeutic Monoclonal Antibodies
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(3), 6565-6565 (2016)
Monosaccharide Analysis
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(1), 6616-6616 (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service