Skip to Content
Merck

Skip To

362000

N-Glycolylneuraminic Acid

Formed by the action of CMP-Neu5Ac hydroxylase. Humans are devoid of this compound due to a mutation in the human CMP-Neu5Ac hydroxylase gene that occurred after the Homo-Pan divergence.

Synonym(s):

N-Glycolylneuraminic Acid, NeuGc, Neu5Gc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C11H19NO10
CAS Number:
Molecular Weight:
325.27
UNSPSC Code:
12352201
NACRES:
NA.79
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N([C@@H]([C@@H](O)[C@H](O)[C@H](O)CO)[C@@H](O)C(=O)C(=O)O)C(=O)CO

InChI

1S/C10H17NO10/c12-1-3(14)6(16)7(17)5(11-4(15)2-13)8(18)9(19)10(20)21/h3,5-8,12-14,16-18H,1-2H2,(H,11,15)(H,20,21)/t3-,5+,6-,7-,8-/m1/s1

InChI key

YIICUBSXBNCXHW-BOSCHMPASA-N

General description

Formed by the action of CMP-Neu5Ac hydroxylase. A major component in great ape tissues except brain; however, it is absent in man due to a mutation in the human CMP-Neu5Ac hydroxylase gene that is reported to have occurred after the Homo-Pan divergence.

Other Notes

Chou, H.H., et al. 1998. Proc. Natl. Acad. Sci. USA 95, 11751.
Malykh, Y.N., et al. 1998. Glycoconj. J. 15, 885.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G9793C8271110138
assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥85% (HPLC)

assay

≥98% (HPLC)

solubility

water: 10 mg/mL

solubility

water: soluble 20 mg/mL

solubility

water: 50 mg/mL, clear, colorless

solubility

water: ≥10 mg/mL

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

form

solid

form

powder

form

powder

form

solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

color

off-white

color

white

color

-

color

white


Still not finding the right product?

Explore all of our products under N-Glycolylneuraminic Acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters