Skip to Content
Merck
All Photos(1)

Documents

1538505

USP

Pilocarpine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

cis-3-Ethyl-4-(1-methyl-5-imidazolylmethyl)dihydro-2(3H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2
CAS Number:
Molecular Weight:
208.26
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C11H16N2O2/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2/h5,7-8,10H,3-4,6H2,1-2H3/t8-,10-/m0/s1

InChI key

QCHFTSOMWOSFHM-WPRPVWTQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pilocarpine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jui-Yang Lai
Drug design, development and therapy, 7, 1273-1285 (2013-11-05)
Ocular drug delivery is one of the most commonly used treatment modalities in the management of glaucoma. We have recently proposed the use of gelatin and poly(N-isopropylacrylamide) (PNIPAAm) graft copolymers as biodegradable in situ forming delivery systems for the intracameral
Yan Dong et al.
Brain research, 1535, 115-123 (2013-09-03)
Oxidative stress is a mechanism of cell death induced by seizures. Antioxidant compounds have neuroprotective effects due to their ability to inhibit free radical production. Autophagy is a process in which cytoplasmic components such as organelles and proteins are delivered
S Alisha Epps et al.
Epilepsy & behavior : E&B, 29(1), 47-52 (2013-08-13)
The bidirectional comorbidity between epilepsy and depression is associated with severe challenges for treatment efficacy and safety, often resulting in poor prognosis and outcome for the patient. We showed previously that rats selectively bred for depression-like behaviors (SwLo rats) also
Jing Li et al.
International journal of pharmaceutics, 455(1-2), 75-84 (2013-08-07)
Poor corneal penetration and short preocular retention of a clinical hydrophilic drug, pilocarpine nitrate (PN), for the treatment of open-angle glaucoma and acute angle-closure glaucoma, limit its ocular application. The purpose of this study was to investigate the potential of
Doodipala Samba Reddy et al.
International journal of molecular sciences, 14(9), 18284-18318 (2013-09-10)
This article describes current experimental models of status epilepticus (SE) and neuronal injury for use in the screening of new therapeutic agents. Epilepsy is a common neurological disorder characterized by recurrent unprovoked seizures. SE is an emergency condition associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service