Skip to Content
Merck
All Photos(1)

Key Documents

PHR1493

Supelco

Pilocarpine Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Pilocarpine hydrochloride, (3S,4R)-4,5-Dihydro-3-ethyl-4-(1-methyl-1H-imidazol-5-ylmethyl)-2(3H)-furanone hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$158.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$158.00

About This Item

Empirical Formula (Hill Notation):
C11H16N2O2 · HCl
CAS Number:
Molecular Weight:
244.72
Beilstein:
4034491
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. P1650000
traceable to USP 1538902

API family

pilocarpine

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

202-205 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CC[C@H]1[C@H](COC1=O)Cc2cncn2C

InChI

1S/C11H16N2O2.ClH/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;/h5,7-8,10H,3-4,6H2,1-2H3;1H/t8-,10-;/m0./s1

InChI key

RNAICSBVACLLGM-GNAZCLTHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Pilocarpine Hydrochloride may be used as a pharmaceutical reference standard for the determination of pilocarpine in opthalmic dispersions[1] and serum samples[2] by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Nonselective muscarinic acetylcholine receptor agonist; used to produce an experimental model of epilepsy.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3041 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of pilocarpine by HPLC in presence of isopilocarpine a pH-sensitive polymer in ophthalmic dispersions
Ibrahim H, et al.
Journal of Pharmaceutical and Biomedical Analysis, 5(4), 379-382 (1987)
Simple chemiluminescence determination of pilocarpine in pharmaceuticals and human serum.
Keyvanfard M, et al.
Acta Chimica Slovenica, 58(3), 379-382 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service