Skip to Content
Merck
All Photos(1)

Key Documents

1533250

USP

Phenylethyl alcohol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Phenylethanol, β-PEA, 2-Phenylethyl alcohol, Benzyl carbinol, PEA

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$544.00

$544.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$544.00

About This Item

Linear Formula:
C6H5CH2CH2OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1905732
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$544.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

vapor density

4.21 (vs air)

vapor pressure

1 mmHg ( 58 °C)

API family

phenylethyl alcohol

manufacturer/tradename

USP

refractive index

n20/D 1.5317 (lit.)

bp

219-221 °C/750 mmHg (lit.)

mp

−27 °C (lit.)

density

1.020 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OCCc1ccccc1

InChI

1S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

WRMNZCZEMHIOCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenylethyl alcohol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fluticasone Propionate Nasal Spray

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service