Skip to Content
Merck

Skip To

1397006

USP

Methacycline hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

6-Demethyl-6-deoxy-6-methyleneoxytetracycline monohydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Check Cart for Availability
$576.00

About This Item

Empirical Formula (Hill Notation):
C22H22N2O8 · HCl
CAS Number:
Molecular Weight:
478.88
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5470951

$576.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

methacycline

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

Cl[H].[H][C@@]12[C@@H](O)[C@]3([H])C(=C)c4cccc(O)c4C(=O)C3=C(O)[C@]1(O)C(=O)C(C(N)=O)=C(O)[C@H]2N(C)C

InChI

1S/C22H22N2O8.ClH/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;/h4-6,10,14-15,17,25,27-29,32H,1H2,2-3H3,(H2,23,31);1H/t10-,14-,15+,17+,22+;/m1./s1

InChI key

VZQARNDJLLWXGL-CCHMMTNSSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methacycline hydrochloride USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Doxycycline
  • Doxycycline Hyclate Capsules
  • Doxycycline Hyclate
  • Doxycycline for Injection
  • Doxycycline Capsules
  • Doxycycline Tablets
  • Doxycycline for Oral Suspension

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
31741PHR1594T8032
grade

pharmaceutical primary standard

grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

format

neat

format

neat

format

neat

format

-

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

−20°C

API family

methacycline

API family

-

API family

methacycline

API family

-


Still not finding the right product?

Explore all of our products under Methacycline hydrochloride


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters