Skip to Content
Merck
Methacycline hydrochloride United States Pharmacopeia (USP) Reference Standard

Methacycline hydrochloride

Methacycline hydrochloride United States Pharmacopeia (USP) Reference Standard
Synonym(s):
6-Demethyl-6-deoxy-6-methyleneoxytetracycline monohydrochloride
Empirical Formula (Hill Notation):
C22H22N2O8 · HCl
CAS Number:
Molecular Weight:
478.88

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1