Skip to Content
Merck
All Photos(1)

Key Documents

1314009

USP

Hydrochlorothiazide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

6-Chloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$544.00

$544.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
$544.00

About This Item

Empirical Formula (Hill Notation):
C7H8ClN3O4S2
CAS Number:
Molecular Weight:
297.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$544.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

hydrochlorothiazide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NS(=O)(=O)c1cc2c(NCNS2(=O)=O)cc1Cl

InChI

1S/C7H8ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-2,10-11H,3H2,(H2,9,12,13)

InChI key

JZUFKLXOESDKRF-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydrochlorothiazide USP reference standard for use in specified quality tests and assays.

Also used to prepare standard, standard stock, system suitability, Hydrochlorothiazide standard solutions for assay, performance, and impurity analysis according to given below monographs of the United States Pharmacopeia (USP):
  • Hydrochlorothiazide Capsules[1]
  • Amiloride Hydrochloride and Hydrochlorothiazide Tablets[2]
  • Captopril and Hydrochlorothiazide Tablets[3]

Biochem/physiol Actions

Carbonic anhydrase inhibitor

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Captopril and Hydrochlorothiazide Tablets
United States Pharmacopeia, 43(4), 737-737 (2019)
Hydrochlorothiazide Capsules
United States Pharmacopeia, 44(1), 2219-2219 (2019)
Amiloride Hydrochloride and Hydrochlorothiazide Tablets
United States Pharmacopeia, 44(3), 224-224 (2019)
Franz H Messerli et al.
The American journal of medicine, 124(10), 896-899 (2011-10-04)
Hydrochlorothiazide (HCTZ) has become by far the most commonly prescribed antihypertensive drug in the US. In 2008, 47.8 million prescriptions were written for HCTZ alone and 87.1 million prescriptions for HCTZ combinations. However, there is no evidence that HCTZ in
Soham Rej et al.
Drugs & aging, 31(12), 911-916 (2014-10-22)
Little is known about how lithium should be dosed to achieve therapeutic but safe serum concentrations in older adults. In this paper, we investigate how the lithium dose-concentration ratio changes across the lifespan. This was a cross-sectional analysis of 63

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service