Skip to Content
Merck
All Photos(1)

Key Documents

PHR1192

Supelco

Benzothiadiazine Related Compound A

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

3-Chloroaniline-4,6-disulfonamide, 4-Amino-6-chloro-1,3-benzenedisulfonamide, 4-Amino-6-chlorobenzene-1,3-disulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$228.00

$228.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
500 MG
$228.00

About This Item

Linear Formula:
H2NC6H2(Cl)(SO2NH2)2
CAS Number:
Molecular Weight:
285.73
Beilstein:
1083877
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$228.00


In StockDetails


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1057507

API family

hydrochlorothiazide

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

257-261 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Nc1cc(Cl)c(cc1S(N)(=O)=O)S(N)(=O)=O

InChI

1S/C6H8ClN3O4S2/c7-3-1-4(8)6(16(10,13)14)2-5(3)15(9,11)12/h1-2H,8H2,(H2,9,11,12)(H2,10,13,14)

InChI key

IHJCXVZDYSXXFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC4018 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biopharmaceutical studies of thiazide diuretics. III. In vivo formation of 2-amino-4-chloro-m-benzenedisulfonamide as a metabolite of hydrochlorothiazide in a patient.
T Okuda et al.
Chemical & pharmaceutical bulletin, 35(8), 3516-3518 (1987-08-01)
M Yamazaki et al.
Chemical & pharmaceutical bulletin, 38(10), 2882-2883 (1990-10-01)
2-Amino-4-chloro-m-benzenedisulfonamide (ACBS) is a metabolite of hydrochlorothiazide. We reported that the ACBS concentration in erythrocytes was higher than in plasma in a patient. Therefore the binding of ACBS to rabbit erythrocyte was studied. The Scatchard plot showed the nonlinear plot
B R Thomas et al.
Journal of chromatography. B, Biomedical applications, 657(2), 383-394 (1994-07-15)
A stability-indicating, quality control analysis method was developed and validated for the diuretic drug substances hydrochlorothiazide (HCTZ) and chlorothiazide (CTZ). Micellar electrokinetic capillary chromatography employing the anionic detergent sodium dodecyl sulfate at 30 mM in 20 mM sodium borate buffer
K Deventer et al.
Journal of chromatography. A, 1216(12), 2466-2473 (2009-02-04)
In sports, thiazide diuretics are used to flush out previously taken prohibited substances with forced diuresis and in sports where weight classes are involved to achieve acute weight loss. Thiazide diuretics include compounds which are very unstable and hydrolyse in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service