1269163
USP
Famciclovir System Suitability Mixture
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Mixture of famciclovir, propionyl famciclovir and 6-chloro famciclovir
Select a Size
Select a Size
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
famciclovir
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
mixture
storage temp.
2-8°C
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Muta. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Following the new USP monograph for Famciclovir Tablets, Purospher™ STAR RP-8 endcapped HPLC columns can be used to monitor organic impurities in Famciclovir tablet formulations. Famciclovir is an antiviral drug indicated for the treatment of herpes zoster, herpes simplex virus 2 (genital herpes), herpes labialis (cold sores), etc.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service