Skip to Content
Merck
All Photos(1)

Key Documents

1051500

USP

Benzethonium chloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(Diisobutylphenoxyethoxyethyl)dimethylbenzylammonium chloride, Phemerol chloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$544.00

$544.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$544.00

About This Item

Empirical Formula (Hill Notation):
C27H42ClNO2
CAS Number:
Molecular Weight:
448.08
Beilstein:
3898548
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$544.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

benzethonium chloride

manufacturer/tradename

USP

mp

162-164 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Cl-].CC(C)(C)CC(C)(C)c1ccc(OCCOCC[N+](C)(C)Cc2ccccc2)cc1

InChI

1S/C27H42NO2.ClH/c1-26(2,3)22-27(4,5)24-13-15-25(16-14-24)30-20-19-29-18-17-28(6,7)21-23-11-9-8-10-12-23;/h8-16H,17-22H2,1-7H3;1H/q+1;/p-1

InChI key

UREZNYTWGJKWBI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Ph

Application

Benzethonium chloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Benzethonium Chloride Topical Solution
  • Methylbenzethonium Chloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

This product is part of USP Biologics Program.
Sales restrictions may apply.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service