Skip to Content
Merck
All Photos(1)

Key Documents

PHR1371

Supelco

Benzalkonium Chloride solution

10%, Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Benzalkonium chloride solution, Alkylbenzyldimethylammonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$177.00

$177.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
5 ML
$177.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$177.00


In StockDetails


Request a Bulk Order

Product Name

Benzalkonium Chloride 10% Solution, Pharmaceutical Secondary Standard; Certified Reference Material

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1051001

API family

benzalkonium chloride

CofA

current certificate can be downloaded

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-30°C

SMILES string

[Cl-].[N+](CCCCCCCCCC)(Cc1ccccc1)(C)C

InChI

1S/C19H34N.ClH/c1-4-5-6-7-8-9-10-14-17-20(2,3)18-19-15-12-11-13-16-19;/h11-13,15-16H,4-10,14,17-18H2,1-3H3;1H/q+1;/p-1

InChI key

TTZLKXKJIMOHHG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2017 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. is a solution 10% w/w?

    1 answer
    1. This product is 10% (w/v). Please view the Sample Certificate of Analysis below for more information:
      https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/rtc/PHR1371_LRAD1895.pdf.

      Helpful?

  4. Can I get additional information regarding the Ratio Determination for C18? Is this standard appropriate for use in a pharmaceutical quality control laboratory? Ar the CoA results provided acceptable fo use in a pharmaceutical QCC lab related to M300?

    1 answer
    1. Methods of determination are provided on the Certificate of Analysis and are according to current compendial monographs. These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available. This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis. The Quality designation of M300 is given according to our M-Clarity™ Program. M300 level products are for products used in applications requiring enhanced change control and quality agreements. For more information on the M-Clarity™ Program, visit the following link: https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management/m-clarity-program

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service