1019428
USP
Amifostine thiol
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
WR-1065, 2-[(3-Aminopropyl)amino]ethanethiol dihydrochloride, WR-1065 dihydrochloride
Select a Size
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
amifostine
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
−20°C
SMILES string
Cl.Cl.NCCCNCCS
InChI
1S/C5H14N2S.2ClH/c6-2-1-3-7-4-5-8;;/h7-8H,1-6H2;2*1H
InChI key
XDRLRDHLCIFZIW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Amifostine
- Amifostine for Injection
Biochem/physiol Actions
Analysis Note
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service