Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 200 mg | Check Cart for Availability | $576.00 |
About This Item
grade
pharmaceutical primary standard
API family
acamprosate
mol wt
monograph mol wt. 400.48 (C10H20CaN2O8S2)
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
CC(=O)NCCCS(=O)(=O)O[Ca]OS(=O)(=O)CCCNC(C)=O
InChI
1S/2C5H11NO4S.Ca/c2*1-5(7)6-3-2-4-11(8,9)10;/h2*2-4H2,1H3,(H,6,7)(H,8,9,10);/q;;+2/p-2
InChI key
BUVGWDNTAWHSKI-UHFFFAOYSA-L
Gene Information
human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879), GRIN1(2902), GRIN2A(2903), GRIN2B(2904), GRIN2C(2905), GRIN2D(2906), GRIN3A(116443), GRIN3B(116444)
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Biochem/physiol Actions
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename BP | manufacturer/tradename - | manufacturer/tradename - |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade - | grade certified reference material, pharmaceutical secondary standard |
| format neat | format neat | format - | format - |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) - | application(s) pharmaceutical small molecule |
| API family acamprosate | API family acamprosate | API family - | API family - |
| mol wt monograph mol wt. 400.48 (C10H20CaN2O8S2) | mol wt - | mol wt - | mol wt - |
Still not finding the right product?
Explore all of our products under Acamprosate calcium
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1000554-200MG | 04061833838907 |
