Skip to Content
Merck
All Photos(1)

Key Documents

1000420

USP

Abacavir Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(1S,4R)-4-[2,6-Diamino-9H-purin-9-yl]-2-cyclopentene-1-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N6O
CAS Number:
Molecular Weight:
246.27
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

abacavir

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[n]2(c3nc(nc(c3nc2)N)N)[C@@H]1C[C@@H](C=C1)CO

InChI

1S/C11H14N6O/c12-9-8-10(16-11(13)15-9)17(5-14-8)7-2-1-6(3-7)4-18/h1-2,5-7,18H,3-4H2,(H4,12,13,15,16)/t6-,7+/m1/s1

InChI key

ZKJUXHDSLJYKED-RQJHMYQMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Abacavir Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Wen et al.
Drug metabolism and disposition: the biological fate of chemicals, 27(1), 113-121 (1999-01-12)
Several in vitro and in situ approaches were used to determine the dominant presystemic activation site for (-)-6-aminocarbovir, (-)-carbocyclic 2',3'-didehydro-2', 3'-dideoxy-6-deoxy-6-aminoguanosine (6AC) conversion in rats. The in vitro disappearance half-lives (mean +/- S.D.) in the cytosolic fractions obtained from homogenates
R L Yeager et al.
Drug metabolism and disposition: the biological fate of chemicals, 19(2), 462-466 (1991-03-01)
The potential for the metabolic conversion of (-)-6-aminocarbovir to (-)-carbovir, a potent reverse transcriptase inhibitor effective against human immunodeficiency virus, has been examined in male Sprague-Dawley rats. Plasma (-)-6-aminocarbovir concentrations declined rapidly in a biphasic manner following an iv bolus
Y D Wen et al.
Pharmaceutical research, 12(6), 911-915 (1995-06-01)
Evaluate the ability of (-)-6-aminocarbovir ((-)-6AC) to improve the CNS exposure to (-)-carbovir ((-)-CBV). Activation of (-)-6AC in vitro was assessed by incubations of rat brain tissue homogenates. The in vivo brain exposure to (-)-CBV was then examined in rats
Evaluation of gastrointestinal absorption and metabolism.
C L Zimmerman et al.
Drug metabolism reviews, 29(4), 957-975 (1998-01-09)
C L Zimmerman et al.
Drug metabolism and disposition: the biological fate of chemicals, 20(1), 47-51 (1992-01-01)
The recently synthesized carbocyclic 2',3'-didehydro-2',3'-dideoxy-6-deoxy-6-amino-guanosine [(-)6AC] was evaluated as a prodrug for carbovir, carbocyclic 2',3'-didehydro-2',3'-dideoxyguanosine [(-)CBV] in seven male Sprague-Dawley rats. A randomized three-way cross-over design was used. Rats were assigned to receive the following treatments: a 20 mg/kg (-)6AC

Related Content

Small molecules are ions and compounds of molecular weight typically less than 900 daltons. These compounds can be effectively separated and analyzed by HPLC, UHPLC and LC-MS using mainly silica particles or monolithic stationary phases with a broad range of column chemistries (modifications).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service