Skip to Content
Merck
All Photos(1)

Key Documents

A6112

Sigma-Aldrich

Calmodulin-Agarose

Calmodulin-Agarose
1 of 1 reviewers received a sample product or took part in a promotion

saline suspension

Synonym(s):

Calmodulin−Agarose, Phosphodiesterase 3′,5′-Cyclic Nucleotide Activator-Agarose from bovine testes

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$236.00
5 ML
$705.00
10 ML
$1,290.00

$236.00


Available to ship onApril 01, 2025Details



Select a Size

Change View
1 ML
$236.00
5 ML
$705.00
10 ML
$1,290.00

About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

$236.00


Available to ship onApril 01, 2025Details


biological source

Saccharomyces cerevisiae

Quality Level

form

saline suspension

extent of labeling

≥0.5 mg per mL

technique(s)

affinity chromatography: suitable

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

suitability

suitable for chromatography

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Calmodulin-agarose is used in affinity chromatography, protein chromatography, calcium associated proteins, and specialty resins. Calmodulin-agarose has been used to suggest a coordinated regulation of processes in both mitochondria and chloroplasts by calcium signals. Calmodulin-agarose has also been used to study apoptosis in prostate cancer cells and to analyze the interaction of the influenza virus polymerase complex with human cell factors.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ninghai Gan et al.
Nature, 572(7769), 387-391 (2019-07-23)
The bacterial pathogen Legionella pneumophila creates an intracellular niche permissive for its replication by extensively modulating host-cell functions using hundreds of effector proteins delivered by its Dot/Icm secretion system1. Among these, members of the SidE family (SidEs) regulate several cellular

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Brighton, United Kingdom
    • Reviews 5
    • Votes 4
    5 out of 5 stars.

    Will thoroughly recommend.

    PI have previously used this reagent. I have obtained similar results this time around.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service