Y0002267
Ethanolamine
CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine
About This Item
Recommended Products
vapor density
2.1 (vs air)
vapor pressure
0.2 mmHg ( 20 °C)
API family
ethanolamine
autoignition temp.
1436 °F
expl. lim.
17 %
manufacturer/tradename
EDQM
refractive index
n20/D 1.454 (lit.)
bp
170 °C (lit.)
69-70 °C/10 mmHg
mp
10-11 °C (lit.)
density
1.012 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
NCCO
InChI
1S/C2H7NO/c3-1-2-4/h4H,1-3H2
InChI key
HZAXFHJVJLSVMW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
195.8 °F - Pensky-Martens closed cup
Flash Point(C)
91 °C - Pensky-Martens closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service