Skip to Content
Merck
All Photos(1)

Key Documents

PHL83911

10-Shogaol

phyproof® Reference Substance

Synonym(s):

[10]-Shogaol, 1-(4-Hydroxy-3-methoxyphenyl)-4-tetradecen-3-one, 10-Shogaol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$515.00

$515.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$515.00

About This Item

Empirical Formula (Hill Notation):
C21H32O3
CAS Number:
Molecular Weight:
332.48
Beilstein:
20347696
UNSPSC Code:
41116107
NACRES:
NA.24

$515.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

−20°C

SMILES string

O=C(/C=C\CCCCCCCCC)CCC1=CC=C(O)C(OC)=C1

InChI

1S/C21H32O3/c1-3-4-5-6-7-8-9-10-11-12-19(22)15-13-18-14-16-20(23)21(17-18)24-2/h11-12,14,16-17,23H,3-10,13,15H2,1-2H3/b12-11-

InChI key

FADFGCOCHHNRHF-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: zingiber

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service