Skip to Content
Merck
All Photos(1)

Key Documents

G-027

Supelco

Ginger Gingerols and Shogaols Mix solution

certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Gingerols/Shogaols Mix Solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$785.00

$785.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$785.00

About This Item

Empirical Formula (Hill Notation):
C17H26O4
Molecular Weight:
294.39
UNSPSC Code:
41116107
NACRES:
NA.24

$785.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

6-Gingerol, 500 μg/mL
8-Gingerol, 500 μg/mL
10-Gingerol, 500 μg/mL
6-Shogaol, 500 μg/mL
8-Shogaol, 500 μg/mL
10-Shogaol, 500 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

format

multi-component solution

storage temp.

−20°C

General description

This phytochemical certified reference materials mix is produced at the Millipore Sigma Cerilliant site in Round Rock Texas under ISO/IEC 17025 and ISO 17034 accreditation. A comprehensive certificate including traceable values and expanded uncertainties for all the analytes is available.

genus zingiber





Discover the entire range of phytochemical certified reference material solutions from Cerilliant

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service