Skip to Content
Merck
All Photos(1)

Documents

PHL80166

Tyrosol

phyproof® Reference Substance

Synonym(s):

2-(4-Hydroxyphenyl)ethanol, 4-Hydroxyphenethyl alcohol, Tyrosol, p-HPEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
1859884
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

mp

89-92 °C (lit.)

storage temp.

2-8°C

SMILES string

OCCc1ccc(O)cc1

InChI

1S/C8H10O2/c9-6-5-7-1-3-8(10)4-2-7/h1-4,9-10H,5-6H2

InChI key

YCCILVSKPBXVIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Antioxidant found in olive oil.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service