Skip to Content
Merck
All Photos(1)

Key Documents

94770

Sigma-Aldrich

Vanillic acid

purum, ≥97.0% (HPLC)

Synonym(s):

4-Hydroxy-3-methoxybenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$75.40
50 G
$251.00
250 G
$778.00

$75.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$75.40
50 G
$251.00
250 G
$778.00

About This Item

Linear Formula:
HOC6H3(OCH3)CO2H
CAS Number:
Molecular Weight:
168.15
Beilstein:
2208364
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$75.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

purum

Quality Level

Assay

≥97.0% (HPLC)

form

powder

ign. residue

≤0.3%

mp

208-210 °C (lit.)
208-213 °C

functional group

carboxylic acid

SMILES string

COc1cc(ccc1O)C(O)=O

InChI

1S/C8H8O4/c1-12-7-4-5(8(10)11)2-3-6(7)9/h2-4,9H,1H3,(H,10,11)

InChI key

WKOLLVMJNQIZCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Vanillic acid is used as a precursor in the synthesis of a wide range of bio-based epoxy resins and polyesters.[1][2][3] It is also a key starting material in the total synthesis of barmumycin[4]4 and bosutinib.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation, structural assignment, and total synthesis of barmumycin.
Lorente A, et al.
The Journal of Organic Chemistry, 75(24), 8508-8515 (2010)
Aromatic thermotropic polyesters based on 2, 5-furandicarboxylic acid and vanillic acid.
Wilsens C H, et al.
Polymer, 55(10), 2432-2439 (2014)
Xiao Jia Yin et al.
Molecules (Basel, Switzerland), 15(6), 4261-4266 (2010-07-27)
This paper reports a novel synthesis of bosutinib starting from 3-methoxy-4-hydroxybenzoic acid. The process starts with esterification of the starting material, followed by alkylation, nitration, reduction, cyclization, chlorination and two successive amination reactions. The intermediates and target molecule were characterized
Chemo-enzymatic functionalization of gallic and vanillic acids: synthesis of bio-based epoxy resins prepolymers.
Aouf C, et al.
Green Chemistry, 14(8), 2328-2336 (2012)
Novel vanillic acid-based poly (ether?ester) s: from synthesis to properties.
Pang C, et al.
Polym. Chem., 6(5), 797-804 (2015)

Protocols

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service